Ethyl (3-Methoxybenzoyl)acetate ≥98.0% (by GC, titration analysis)
Supplier: TCI America
E1121-1G
E1121-5G
TCE1121-1GEA
97.49
CAD
TCE1121-1G
TCE1121-5G
Ethyl (3-Methoxybenzoyl)acetate ≥98.0% (by GC, titration analysis)
Ethyl (3-Methoxybenzoyl)acetate
Formula:
C₁₂H₁₄O₄ Boiling Pt: 123°C |
MDL Number:
MFCD00086704 CAS Number: 27834-99-7 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % (total of isomer) |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...