1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D5064-1G D5064-5G
76219-820EA 83.74 CAD
76219-820 76219-822
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone ≥97.0% (by GC, titration analysis)
1-(3,4-Dimethylphenyl)-3-methyl-5-pyrazolone
Formula: C₁₂H₁₄N₂O
MDL Number: MFCD09795980
CAS Number: 277299-70-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR