1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

T3653-100G T3653-500G
76220-700EA 179.91 CAD
76220-700 76220-702
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione ≥98.0% (by HPLC, total nitrogen)
1,3,5-Tris(3,5-di-tert-butyl-4-hydroxybenzyl)-1,3,5-triazinane-2,4,6-trione
Formula: C₄₈H₆₉N₃O₆
MDL Number: MFCD00134700
CAS Number: 27676-62-6
EINECS: 248-597-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 220.0 to 224.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR