trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
(E)-3-(4-Acetoxyphenyl)acrylic acid, trans-3-(4-Acetoxyphenyl)-2-propenoic acid
A2845-25G
A2845-5G
TCA2845-25GEA
799.48
CAD
TCA2845-25G
TCA2845-5G
trans-4-Acetoxycinnamic acid ≥98.0% (by GC, titration analysis)
trans-4-Acetoxycinnamic acid
Formula:
C₁₁H₁₀O₄ |
MDL Number:
MFCD00016847 CAS Number: 27542-85-4 EINECS: 239-512-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 208.0 to 212.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...