7-Ethynylcoumarin ≥98.0% (by GC)
Supplier: TCI America
E1092-1G
E1092-200MG
TCE1092-1GEA
761.83
CAD
TCE1092-1G
TCE1092-200MG
7-Ethynylcoumarin ≥98.0% (by GC)
7-Ethynylcoumarin
Formula:
C₁₁H₆O₂ |
CAS Number:
270088-04-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 132.0 to 136.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...