3'-Bromo-2'-methoxyacetophenone ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5498-1G
B5498-200MG
TCB5498-1GEA
477.16
CAD
TCB5498-1G
TCB5498-200MG
3'-Bromo-2'-methoxyacetophenone ≥98.0% (by GC)
3'-Bromo-2'-methoxyacetophenone
Formula:
C₉H₉BrO₂ |
MDL Number:
MFCD22573042 CAS Number: 267651-23-0 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...