3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Pentaerythritol bis(2,4-di-tert-butylphenylphosphite)

B5664-25G B5664-5G
TCB5664-25GEA 199.08 CAD
TCB5664-25G TCB5664-5G
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane ≥95.0% (by GC)
3,9-Bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Formula: C₃₃H₅₀O₆P₂
MW: 604.70 g/mol
MDL Number: MFCD00071521
CAS Number: 26741-53-7
EINECS: 247-952-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR