4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl ≥95.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)-1,1'-biphenyl

B4977-1G B4977-200MG
TCB4977-1GEA 1031.74 CAD
TCB4977-1G TCB4977-200MG
4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl ≥95.0% (by HPLC, total nitrogen)
4,4'-Bis(4,6-diphenyl-1,3,5-triazin-2-yl)biphenyl
Formula: C₄₂H₂₈N₆
MW: 616.73 g/mol
MDL Number: MFCD28291963
CAS Number: 266349-83-1

Order Now

Specification Test Results

Appearance White to Light gray to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR