4-(2-Propynyloxy)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

P2224-1G P2224-5G
TCP2224-1GEA 383.48 CAD
TCP2224-1G TCP2224-5G
4-(2-Propynyloxy)aniline ≥98.0% (by GC, titration analysis)
4-(2-Propynyloxy)aniline
Formula: C₉H₉NO
Boiling Pt: 95°C
Storage Temperature: Refrigerator
MDL Number: MFCD12823190
CAS Number: 26557-78-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR