1-Azaphenothiazine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A2862-1G A2862-200MG
TCA2862-1GEA 370.19 CAD
TCA2862-1G TCA2862-200MG
1-Azaphenothiazine ≥98.0% (by GC, titration analysis)
1-Azaphenothiazine
Formula: C₁₁H₈N₂S
Boiling Pt: 370°C
MDL Number: MFCD00160449
CAS Number: 261-96-1
EINECS: 205-973-7

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR