3,6-Dibromophthalic anhydride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
4,7-Dibromoisobenzofuran-1,3-dione
D4724-1G
D4724-200MG
TCD4724-1GEA
788.42
CAD
TCD4724-1G
TCD4724-200MG
3,6-Dibromophthalic anhydride ≥98.0% (by titrimetric analysis)
3,6-Dibromophthalic anhydride
Formula:
C₈H₂Br₂O₃ MW: 305.91 g/mol |
MDL Number:
MFCD01117424 CAS Number: 25834-16-6 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 95.0 % |
Purity (Methanolysis Method) | min. 98.0 % |
Melting point | 207.0 to 212.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...