3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine ≥95.0% (by GC)
Supplier: TCI America
D5423-1G
D5423-200MG
TCD5423-1GEA
499.28
CAD
TCD5423-1G
TCD5423-200MG
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine ≥95.0% (by GC)
3,3-Dimethyl-3,4-dihydro-2H-thieno[3,4-b][1,4]dioxepine
Formula:
C₉H₁₂O₂S Storage Temperature: Refrigerator |
MDL Number:
MFCD08705268 CAS Number: 255901-50-9 |
Specification Test Results
Appearance | White to Yellow to Green powder to lump |
Purity (GC) | min. 95.0 % |
Melting point | 42.0 to 46.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...