[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
Boron, [2-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)(4-iodophenyl)methyl]-3,5-dimethyl-1H-pyrrolato-κN]difluoro-, (T-4)-
D4632-1G
D4632-200MG
76219-048EA
941.68
CAD
76219-048
76219-050
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) ≥98.0% (by HPLC)
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane)
Formula:
C₁₉H₁₈BF₂IN₂ |
MDL Number:
MFCD29905036 CAS Number: 250734-47-5 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 5.60 to 6.80 % |
Lambda max. | 500.0 to 505.0 nm (CH2Cl2) |
Molar extinction coefficient | min. 88000 (CH2Cl2, 500.0 to 505.0nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...