n-Octyl acrylate ≥98.0% (by GC) stabilized

Supplier: TCI America
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

O0478-25ML O0478-500ML
TCO0478-25MLEA 79.4 CAD
TCO0478-25ML TCO0478-500ML
n-Octyl acrylate ≥98.0% (by GC) stabilized
n-Octyl acrylate
Formula: C₁₁H₂₀O₂
Boiling Pt: 57°C
MDL Number: MFCD00048935
CAS Number: 2499-59-4
EINECS: 219-696-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR