Benzo[b]benzo[4,5]thieno[2,3-d]thiophene ≥97.0% (by GC)

Supplier: TCI America

B5551-1G B5551-200MG
TCB5551-1GEA 597.24 CAD
TCB5551-1G TCB5551-200MG
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene ≥97.0% (by GC)
Benzo[b]benzo[4,5]thieno[2,3-d]thiophene
Formula: C₁₄H₈S₂
MDL Number: MFCD02114004
CAS Number: 248-70-4

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 218.0 to 222.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR