Dibromopyromellitic Dianhydride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 4,8-Dibromobenzo[1,2-c:4,5-c']difuran-1,3,5,7-tetraone, 3,6-Dibromobenzene-1,2,4,5-tetracarboxylic 1,2:4,5-Dianhydride

D4947-1G D4947-5G
TCD4947-1GEA 474 CAD
TCD4947-1G TCD4947-5G
Dibromopyromellitic Dianhydride ≥98.0% (by HPLC)
Dibromopyromellitic Dianhydride
Formula: C₁₀Br₂O₆
MW: 375.91 g/mol
Melting Pt: 275°C
MDL Number: MFCD01100205
CAS Number: 24848-78-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Methanolysis Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR