1-Methoxy-4-methylnaphthalene ≥97.0% (by GC)
Supplier: TCI America
M2930-1G
M2930-200MG
76220-308EA
230.68
CAD
76220-308
76220-310
1-Methoxy-4-methylnaphthalene ≥97.0% (by GC)
1-Methoxy-4-methylnaphthalene
Formula:
C₁₂H₁₂O Boiling Pt: 155°C |
MDL Number:
MFCD00021472 CAS Number: 24821-54-3 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...