3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide) ≥98.0% (by total nitrogen basis)
Supplier: TCI America
|
Warning
|
M2790-1G
M2790-5G
TCM2790-1GEA
156.42
CAD
TCM2790-1G
TCM2790-5G
3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide) ≥98.0% (by total nitrogen basis)
3,3'-Methylenebis(1-tert-butyl-3-imidazolium bromide)
Formula:
C₁₅H₂₆Br₂N₄ |
CAS Number:
247902-48-3 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 96.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...