2-Decylthiophene ≥97.0% (by GC)

Supplier: TCI America

D5240-1ML D5240-5ML
TCD5240-1MLEA 112.18 CAD
TCD5240-1ML TCD5240-5ML
2-Decylthiophene ≥97.0% (by GC)
2-Decylthiophene
Formula: C₁₄H₂₄S
MW: 224.41 g/mol
Boiling Pt: 167°C
MDL Number: MFCD00715761
CAS Number: 24769-39-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR