9,10-Bis(4-methoxyphenyl)anthracene ≥98.0% (by GC)

Supplier: TCI America

B5022-1G B5022-200MG
TCB5022-1GEA 475.58 CAD
TCB5022-1G TCB5022-200MG
9,10-Bis(4-methoxyphenyl)anthracene ≥98.0% (by GC)
9,10-Bis(4-methoxyphenyl)anthracene
Formula: C₂₈H₂₂O₂
MDL Number: MFCD00009936
CAS Number: 24672-76-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR