5H-Pyrido[4,3-b]indole ≥98.0% (by GC)

Supplier: TCI America

Synonyms: gamma-Carboline

P2362-1G P2362-200MG
TCP2362-1GEA 545.1 CAD
TCP2362-1G TCP2362-200MG
5H-Pyrido[4,3-b]indole ≥98.0% (by GC)
5H-Pyrido[4,3-b]indole
Formula: C₁₁H₈N₂
MW: 168.20 g/mol
MDL Number: MFCD00965976
CAS Number: 244-69-9
Merck Index: 14,01805

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 231.0 to 235.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR