4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)
Supplier: TCI America
D5281-1G
D5281-200MG
TCD5281-1GEA
714.16
CAD
TCD5281-1G
TCD5281-200MG
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl ≥97.0% (by GC)
4,4'-Dihydroxy-3,3',5,5'-tetraisopropylbiphenyl
Formula:
C₂₄H₃₄O₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD20257818 CAS Number: 2416-95-7 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 107.0 to 111.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...