2-Bromo-3-cyclohexylthiophene ≥95.0% (by GC)

Supplier: TCI America

B5420-1G B5420-5G
TCB5420-1GEA 325.48 CAD
TCB5420-1G TCB5420-5G
2-Bromo-3-cyclohexylthiophene ≥95.0% (by GC)
2-Bromo-3-cyclohexylthiophene
Formula: C₁₀H₁₃BrS
Storage Temperature: Refrigerator
CAS Number: 241477-71-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR