Methyl 3,5-Dihydroxy-4-methoxybenzoate ≥98.0% (by GC)

Supplier: TCI America
Warning

M2794-1G M2794-5G
TCM2794-1GEA 213.3 CAD
TCM2794-1G TCM2794-5G
Methyl 3,5-Dihydroxy-4-methoxybenzoate ≥98.0% (by GC)
Methyl 3,5-Dihydroxy-4-methoxybenzoate
Formula: C₉H₁₀O₅
MDL Number: MFCD00017195
CAS Number: 24093-81-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 147.0 to 151.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR