9,10-Bis(6-methoxy-2-naphthyl)anthracene ≥98.0% (by HPLC)

Supplier: TCI America

B5046-1G B5046-200MG
TCB5046-1GEA 704.68 CAD
TCB5046-1G TCB5046-200MG
9,10-Bis(6-methoxy-2-naphthyl)anthracene ≥98.0% (by HPLC)
9,10-Bis(6-methoxy-2-naphthyl)anthracene
Formula: C₃₆H₂₆O₂
MW: 490.60 g/mol
MDL Number: MFCD12022382
CAS Number: 235099-48-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR