Guanabenz Acetate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

G0456-1G G0456-200MG
TCG0456-1GEA 150.1 CAD
TCG0456-1G TCG0456-200MG
Guanabenz Acetate ≥98.0% (by HPLC, titration analysis)
Guanabenz Acetate
Formula: C₈H₈Cl₂N₄·C₂H₄O₂
MDL Number: MFCD00153801
CAS Number: 23256-50-0
EINECS: 245-534-7
UN: 2811
ADR: 6.1,III
Merck Index: 14,04558

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 188.0 to 192.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR