1,3-Dibromo-5-dodecylbenzene ≥97.0% (by GC)

Supplier: TCI America
Warning

D5274-1G D5274-5G
76219-886EA 203.82 CAD
76219-886 76219-888
1,3-Dibromo-5-dodecylbenzene ≥97.0% (by GC)
1,3-Dibromo-5-dodecylbenzene
Formula: C₁₈H₂₈Br₂
Melting Pt: 53°C
Storage Temperature: Refrigerator
CAS Number: 231606-27-2

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to lump
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR