3-Bromo-2,6-dimethoxytoluene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4745-1G B4745-200MG
TCB4745-1GEA 334.96 CAD
TCB4745-1G TCB4745-200MG
3-Bromo-2,6-dimethoxytoluene ≥97.0% (by GC)
3-Bromo-2,6-dimethoxytoluene
Formula: C₉H₁₁BrO₂
Boiling Pt: 96°C
MDL Number: MFCD11046750
CAS Number: 22794-95-2

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR