2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
Supplier: TCI America
Synonyms:
DMFL-CBP, 9,9'-(9,9-Dimethyl-9H-fluorene-2,7-diyl)bis(9H-carbazole)
B4960-1G
B4960-200MG
76219-468EA
252.8
CAD
76219-468
76219-470
2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene ≥98.0% (by HPLC, total nitrogen)
2,7-Bis(9H-carbazol-9-yl)-9,9-dimethylfluorene
Formula:
C₃₉H₂₈N₂ |
MDL Number:
MFCD12022460 CAS Number: 226958-06-1 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...