2-Bromo-9,9-dihexylfluorene ≥97.0% (by GC)

Supplier: TCI America
Warning

B4404-1G B4404-200MG
TCB4404-1GEA 256.28 CAD
TCB4404-1G TCB4404-200MG
2-Bromo-9,9-dihexylfluorene ≥97.0% (by GC)
2-Bromo-9,9-dihexylfluorene
Formula: C₂₅H₃₃Br
MDL Number: MFCD14584649
CAS Number: 226070-05-9

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid to slightly cloudy liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR