(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
B5244-1G
B5244-5G
76219-542EA
96.38
CAD
76219-542
76219-544
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol ≥98.0% (by GC)
(S)-1-[3,5-Bis(trifluoromethyl)phenyl]ethanol
Formula:
C₁₀H₈F₆O |
MDL Number:
MFCD03093011 CAS Number: 225920-05-8 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Optical purity (GC) | min. 98.0 ee% |
Melting point | 53.0 to 57.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...