2,5-Dichloro-3,4-ethylenedioxythiophene ≥98.0% (by GC)

Supplier: TCI America
Warning

D4746-1G D4746-5G
TCD4746-1GEA 161.95 CAD
TCD4746-1G TCD4746-5G
2,5-Dichloro-3,4-ethylenedioxythiophene ≥98.0% (by GC)
2,5-Dichloro-3,4-ethylenedioxythiophene
Formula: C₆H₄Cl₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD09907757
CAS Number: 225518-49-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR