Ethylene Glycol bis(3-mercaptopropionate) ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: Ethylene bis(3-mercaptopropionate), Glycol bis(3-mercaptopropionate)

E1269-25G E1269-500G
TCE1269-25GEA 91.64 CAD
TCE1269-25G TCE1269-500G
Ethylene Glycol bis(3-mercaptopropionate) ≥97.0% (by titrimetric analysis)
Ethylene Glycol bis(3-mercaptopropionate)
Formula: C₈H₁₄O₄S₂
MW: 238.32 g/mol
Flash Pt: 255°C
MDL Number: MFCD00053520
CAS Number: 22504-50-3
EINECS: 245-044-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 92.0 %
Purity (Iodometric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR