Ethyl (3-Hydroxyphenyl)acetate ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
E1242-1G
E1242-5G
TCE1242-1GEA
60.04
CAD
TCE1242-1G
TCE1242-5G
Ethyl (3-Hydroxyphenyl)acetate ≥98.0% (by GC)
Ethyl (3-Hydroxyphenyl)acetate
Formula:
C₁₀H₁₂O₃ Boiling Pt: 179°C |
MDL Number:
MFCD07369387 CAS Number: 22446-38-4 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...