3-(tert-Butyl)-9H-carbazole ≥95.0% (by GC)

Supplier: TCI America
Warning

B5053-1G B5053-200MG
TCB5053-1GEA 347.6 CAD
TCB5053-1G TCB5053-200MG
3-(tert-Butyl)-9H-carbazole ≥95.0% (by GC)
3-(tert-Butyl)-9H-carbazole
Formula: C₁₆H₁₇N
MW: 223.32 g/mol
MDL Number: MFCD12024281
CAS Number: 22401-74-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR