Trimethoxy(pentafluorophenyl)silane ≥97.0% (by GC)

Supplier: TCI America
Warning

T3352-1G T3352-5G
TCT3352-1GEA 206.98 CAD
TCT3352-1G TCT3352-5G
Trimethoxy(pentafluorophenyl)silane ≥97.0% (by GC)
Trimethoxy(pentafluorophenyl)silane
Formula: C₉H₉F₅O₃Si
MDL Number: MFCD04039205
CAS Number: 223668-64-2

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR