2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene ≥98.0% (by HPLC)

Supplier: TCI America

B4926-1G B4926-200MG
76219-448EA 229.1 CAD
76219-448 76219-450
2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene ≥98.0% (by HPLC)
2,7-Bis[N-(1-naphthyl)anilino]-9,9-dimethylfluorene
Formula: C₄₇H₃₆N₂
MDL Number: MFCD11110702
CAS Number: 222319-05-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR