1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene ≥98.0% (by HPLC)
Supplier: TCI America
B5605-1G
B5605-5G
76218-854EA
118.5
CAD
76218-854
76218-856
1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene ≥98.0% (by HPLC)
1,2-Bis[(3,6-dibromo-9H-carbazol-9-yl)methyl]benzene
Formula:
C₃₂H₂₀Br₄N₂ Melting Pt: 319°C |
CAS Number:
222166-46-3 |
Specification Test Results
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 3.30 to 4.10 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...