1-(2-Pyrimidinyl)-1H-indole ≥98.0% (by GC)
Supplier: TCI America
P2505-1G
P2505-5G
76220-450EA
304.94
CAD
76220-450
76220-452
1-(2-Pyrimidinyl)-1H-indole ≥98.0% (by GC)
1-(2-Pyrimidinyl)-1H-indole
Formula:
C₁₂H₉N₃ |
MDL Number:
MFCD28556906 CAS Number: 221044-05-9 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 95.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...