1-Bromo-2-phenylnaphthalene ≥98.0% (by GC)

Supplier: TCI America

B4636-1G B4636-200MG
TCB4636-1GEA 545.42 CAD
TCB4636-1G TCB4636-200MG
1-Bromo-2-phenylnaphthalene ≥98.0% (by GC)
1-Bromo-2-phenylnaphthalene
Formula: C₁₆H₁₁Br
Boiling Pt: 170°C
MDL Number: MFCD28384138
CAS Number: 22082-93-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 63.0 to 67.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR