1-Propene 1,3-Sultone ≥99.0% (by GC)
Supplier: TCI America
|
Warning
|
P2260-25G
P2260-5G
TCP2260-25GEA
492.96
CAD
TCP2260-25G
TCP2260-5G
1-Propene 1,3-Sultone ≥99.0% (by GC)
1-Propene 1,3-Sultone
Formula:
C₃H₄O₃S Boiling Pt: 118°C |
MDL Number:
MFCD12405143 CAS Number: 21806-61-1 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 83.0 to 87.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...