Nalpha-tert-Butoxycarbonyl-Nepsilon-tetradecanoyl-L-lysine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Nalpha-Boc-Nepsilon-tetradecanoyl-L-lysine, (S)-2-((tert-Butoxycarbonyl)amino)-6-tetradecanamidohexanoic acid, Boc-Lys(Myr)-OH

B5366-1G B5366-200MG
TCB5366-1GEA 662.02 CAD
TCB5366-1G TCB5366-200MG
Nalpha-tert-Butoxycarbonyl-Nepsilon-tetradecanoyl-L-lysine ≥97.0% (by titrimetric analysis)
Nalpha-tert-Butoxycarbonyl-Nepsilon-tetradecanoyl-L-lysine
Formula: C₂₅H₄₈N₂O₅
MDL Number: MFCD30725738
CAS Number: 2173052-71-4

Order Now

Specification Test Results

Purity (HPLC) min. 95.0 area%
Purity (Neutralization titration) min. 97.0 %
Melting point 87.0 to 92.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR