1-Acryloyloxy-3-hydroxyadamantane ≥98.0% (by GC)

Supplier: TCI America
Warning

A2859-25G A2859-5G
TCA2859-25GEA 504.6 CAD
TCA2859-25G TCA2859-5G
1-Acryloyloxy-3-hydroxyadamantane ≥98.0% (by GC)
1-Acryloyloxy-3-hydroxyadamantane
Formula: C₁₃H₁₈O₃
MDL Number: MFCD13194929
CAS Number: 216581-76-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR