Cyclohexanemethylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
|
Warning
|
Synonyms:
Cyclohexanemethylammonium iodide, (Aminomethyl)cyclohexane Hydroiodide, Cyclohexylmethylamine Hydroiodide
C3425-1G
C3425-5G
TCC3425-1GEA
102.7
CAD
TCC3425-1G
TCC3425-5G
Cyclohexanemethylamine Hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Cyclohexanemethylamine Hydroiodide
Formula:
C₇H₁₅N·HI MW: 241.12 g/mol |
CAS Number:
2153504-15-3 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (with Total Nitrogen) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...