5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene ≥95.0% (by GC)

Supplier: TCI America

D4184-1G D4184-200MG
TCD4184-1GEA 733.91 CAD
TCD4184-1G TCD4184-200MG
5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene ≥95.0% (by GC)
5,5'-Dibromo-4,4'-dihexyl-2,2'-bithiophene
Formula: C₂₀H₂₈Br₂S₂
Storage Temperature: Refrigerator
CAS Number: 214493-03-5

Order Now

Specification Test Results

Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR