Ethyl 2-Chloro-6-fluorophenylacetate ≥98.0% (by GC)

Supplier: TCI America

E1038-1G E1038-5G
TCE1038-1GEA 161.16 CAD
TCE1038-1G TCE1038-5G
Ethyl 2-Chloro-6-fluorophenylacetate ≥98.0% (by GC)
Ethyl 2-Chloro-6-fluorophenylacetate
Formula: C₁₀H₁₀ClFO₂
MDL Number: MFCD00800606
CAS Number: 214262-85-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR