3-Chloropyrazine-2-carboxamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

C3376-1G C3376-200MG
TCC3376-1GEA 270.18 CAD
TCC3376-1G TCC3376-200MG
3-Chloropyrazine-2-carboxamide ≥98.0% (by HPLC, total nitrogen)
3-Chloropyrazine-2-carboxamide
Formula: C₅H₄ClN₃O
MDL Number: MFCD09863915
CAS Number: 21279-62-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR