(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: (11bS)-N,N-Bis[(1S)-1-phenylethyl]dinaphtho[2,1-d:1',2'-f][1,3,2]dioxaphosphepin-4-amine

D4995-1G D4995-200MG
76219-112EA 722.06 CAD
76219-112 76219-114
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine ≥98.0% (by HPLC)
(S,S,S)-(3,5-Dioxa-4-phosphacyclohepta[2,1-a:3,4-a']dinaphthalen-4-yl)bis(1-phenylethyl)amine
Formula: C₃₆H₃₀NO₂P
MDL Number: MFCD04117688
CAS Number: 209482-27-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 95.0 area%
Optical purity (LC) min. 98.0 ee%
Elemental analysis (Nitrogen) 2.20 to 3.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR