N-Propargylmaleimide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

P2139-100MG P2139-25MG
TCP2139-100MGEA 172.01 CAD
TCP2139-100MG TCP2139-25MG
N-Propargylmaleimide ≥98.0% (by HPLC, total nitrogen)
N-Propargylmaleimide
Formula: C₇H₅NO₂
Boiling Pt: 88°C
MDL Number: MFCD12090919
CAS Number: 209395-32-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 55.0 to 60.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR