Triethylene Glycol Mono(2-propynyl) ether ≥96.0% (by GC)

Supplier: TCI America

Synonyms: Propargyl-PEG3-alcohol, 2-[2-[2-(2-Propynyloxy)ethoxy]ethoxy]ethanol, Triethylene Glycol Monopropargyl ether

T3114-1G T3114-5G
TCT3114-1GEA 145.36 CAD
TCT3114-1G TCT3114-5G
Triethylene Glycol Mono(2-propynyl) ether ≥96.0% (by GC)
Triethylene Glycol Mono(2-propynyl) ether
Formula: C₉H₁₆O₄
MW: 188.22 g/mol
MDL Number: MFCD13189984
CAS Number: 208827-90-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR